5-Bromo-2-chloro-N-cyclohexylnicotinamide ≥97%

Supplier: AOBChem USA

Warning

47425-500MG 47425-1G 47425-5G 47425-25G
77327-346EA 202.91 USD
77327-346 77350-924 77363-690 77386-190
5-Bromo-2-chloro-N-cyclohexylnicotinamide ≥97%
5-Bromo-2-chloro-N-cyclohexylnicotinamide

Precaución: for Research Use Only

Formula: C₁₂H₁₄BrClN₂O
MW: 317.609 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15504347
CAS Number: 1461685-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR