4-Fluoro-3-iodo-6-(methylthio)-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

47406-500MG 47406-1G 47406-5G
77339-478EA 593.28 USD
77339-478 77350-922 77375-256
4-Fluoro-3-iodo-6-(methylthio)-1H-indazole ≥97%
4-Fluoro-3-iodo-6-(methylthio)-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₆FIN₂S
MW: 308.115 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707271
CAS Number: 2484888-76-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR