(E)-methyl 3-(3-fluoro-5-(methylthio)phenyl)acrylate ≥97%

Supplier: AOBChem USA

Warning

47405-500MG 47405-1G 47405-5G 47405-25G
77327-344EA 123.6 USD
77327-344 77344-032 77363-686 77375-254
(E)-methyl 3-(3-fluoro-5-(methylthio)phenyl)acrylate ≥97%
(E)-methyl 3-(3-fluoro-5-(methylthio)phenyl)acrylate

Precaución: for Research Use Only

Formula: C₁₁H₁₁FO₂S
MW: 226.267 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564104
CAS Number: 2734780-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR