N'-(1-(3-bromo-4-fluorophenyl)ethylidene)-4-methylbenzenesulfonohydrazide ≥97%

Supplier: AOBChem USA

Warning

47400-1G 47400-5G 47400-25G
77341-008EA 66.95 USD
77341-008 77363-682 77375-250
N'-(1-(3-bromo-4-fluorophenyl)ethylidene)-4-methylbenzenesulfonohydrazide ≥97%
N'-(1-(3-bromo-4-fluorophenyl)ethylidene)-4-methylbenzenesulfonohydrazide

Precaución: for Research Use Only

Formula: C₁₅H₁₄BrFN₂O₂S
MW: 385.251 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707269
CAS Number: 2565832-35-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR