2-Propoxynicotinic acid ≥97%

Supplier: AOBChem USA

Warning

47369-500MG 47369-1G 47369-5G 47369-25G
77327-342EA 160.68 USD
77327-342 77345-790 77363-680 77386-188
2-Propoxynicotinic acid ≥97%
2-Propoxynicotinic acid

Precaución: for Research Use Only

Formula: C₉H₁₁NO₃
Storage Temperature: Ambient
MDL Number: MFCD11619130
CAS Number: 68359-09-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR