(2-propoxypyridin-3-yl)methanol ≥95%

Supplier: AOBChem USA

Warning

47368-500MG 47368-1G 47368-5G 47368-25G
77327-340EA 155.53 USD
77327-340 77345-588 77363-678 77386-186
(2-propoxypyridin-3-yl)methanol ≥95%
(2-propoxypyridin-3-yl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₃NO₂
MW: 167.205 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14606563
CAS Number: 1248798-32-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR