1-Bromo-4-(1-(4-methoxyphenyl)ethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

47364-500MG 47364-1G 47364-5G
77336-386EA 387.28 USD
77336-386 77350-920 77375-248
1-Bromo-4-(1-(4-methoxyphenyl)ethyl)benzene ≥97%
1-Bromo-4-(1-(4-methoxyphenyl)ethyl)benzene

Precaución: for Research Use Only

Formula: C₁₅H₁₅BrO
MW: 291.183 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707268
CAS Number: 65699-87-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR