1-Chloro-2-(1-(p-tolyl)ethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

47357-500MG 47357-1G 47357-5G
77337-092EA 407.88 USD
77337-092 77350-916 77375-244
1-Chloro-2-(1-(p-tolyl)ethyl)benzene ≥95%
1-Chloro-2-(1-(p-tolyl)ethyl)benzene

Precaución: for Research Use Only

Formula: C₁₅H₁₅Cl
MW: 230.733 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707262
CAS Number: 1190968-76-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR