1-Chloro-3-fluoro-4-(methoxymethoxy)-2-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

47311-500MG 47311-1G 47311-5G 47311-25G
77327-336EA 99.91 USD
77327-336 77342-796 77363-674 77375-238
1-Chloro-3-fluoro-4-(methoxymethoxy)-2-methylbenzene ≥95%
1-Chloro-3-fluoro-4-(methoxymethoxy)-2-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₀ClFO₂
MW: 204.6259 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707259
CAS Number: 2432848-96-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR