2,4-Dichloro-6-hydroxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

47310-500MG 47310-1G 47310-5G 47310-25G
77327-334EA 243.08 USD
77327-334 77350-912 77363-672 77386-184
2,4-Dichloro-6-hydroxybenzoic acid ≥97%
2,4-Dichloro-6-hydroxybenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄Cl₂O₃
MW: 207.011 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11203640
CAS Number: 99725-34-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR