Ethyl 3-fluoro-4-(methylthio)benzoate ≥97%

Supplier: AOBChem USA

Warning

47304-500MG 47304-1G 47304-5G 47304-25G
77327-330EA 225.57 USD
77327-330 77350-906 77363-668 77386-180
Ethyl 3-fluoro-4-(methylthio)benzoate ≥97%
Ethyl 3-fluoro-4-(methylthio)benzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₂S
Storage Temperature: Ambient
MDL Number: MFCD33403851
CAS Number: 2635937-60-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR