3-Fluoro-4-(methylthio)benzoic acid ≥95%

Supplier: AOBChem USA

Warning

47302-500MG 47302-1G 47302-5G 47302-25G
77327-326EA 140.08 USD
77327-326 77344-712 77363-664 77386-176
3-Fluoro-4-(methylthio)benzoic acid ≥95%
3-Fluoro-4-(methylthio)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₇FO₂S
Storage Temperature: Ambient
MDL Number: MFCD16853039
CAS Number: 244606-32-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR