6-Bromo-2-fluoro-3-(methylthio)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

47298-500MG 47298-1G 47298-25G 47298-5G
77327-322EA 285.31 USD
77327-322 77350-900 77386-174 77375-232
6-Bromo-2-fluoro-3-(methylthio)benzoic acid ≥97%
6-Bromo-2-fluoro-3-(methylthio)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₆BrFO₂S
Storage Temperature: Ambient
MDL Number: MFCD34756363
CAS Number: 2901085-83-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR