1-(6-Bromo-2-fluoro-3-(methylthio)phenyl)ethanol ≥97%

Supplier: AOBChem USA

Warning

47295-500MG 47295-1G 47295-5G 47295-25G
77327-320EA 99.91 USD
77327-320 77342-794 77363-662 77375-230
1-(6-Bromo-2-fluoro-3-(methylthio)phenyl)ethanol ≥97%
1-(6-Bromo-2-fluoro-3-(methylthio)phenyl)ethanol

Precaución: for Research Use Only

Formula: C₉H₁₀BrFOS
MW: 265.143 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756361
CAS Number: 2901065-55-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR