5-Butoxy-2-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

47284-500MG 47284-1G 47284-5G 47284-25G
77327-318EA 181.28 USD
77327-318 77346-846 77363-658 77386-172
5-Butoxy-2-methylbenzoic acid ≥97%
5-Butoxy-2-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₃
MW: 208.254 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19252308
CAS Number: 1284869-84-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR