(5-Bromo-2,4-dimethylphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

47277-500MG 47277-1G 47277-5G
77336-922EA 400.67 USD
77336-922 77350-896 77375-224
(5-Bromo-2,4-dimethylphenyl)methanol ≥97%
(5-Bromo-2,4-dimethylphenyl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₁BrO
MW: 215.087 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32664259
CAS Number: 1785359-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR