Ethyl 3-chloro-2-fluoro-5-iodobenzoate ≥95%

Supplier: AOBChem USA

Warning

47270-500MG 47270-1G 47270-5G 47270-25G
77327-314EA 262.65 USD
77327-314 77350-892 77363-654 77386-170
Ethyl 3-chloro-2-fluoro-5-iodobenzoate ≥95%
Ethyl 3-chloro-2-fluoro-5-iodobenzoate

Precaución: for Research Use Only

Formula: C₉H₇ClFIO₂
MW: 328.507 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24107279
CAS Number: 1508828-69-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR