(3-Chloro-2-fluoro-5-iodophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

47265-1G 47265-5G 47265-25G
77342-002EA 111.24 USD
77342-002 77363-648 77375-218
(3-Chloro-2-fluoro-5-iodophenyl)(methyl)sulfane ≥97%
(3-Chloro-2-fluoro-5-iodophenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₇H₅ClFIS
MW: 302.535 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902704
CAS Number: 2586126-51-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR