1-Chloro-2-fluoro-5-iodo-3-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

47264-500MG 47264-1G 47264-5G 47264-25G
77327-308EA 120.51 USD
77327-308 77343-718 77363-646 77375-216
1-Chloro-2-fluoro-5-iodo-3-methylbenzene ≥95%
1-Chloro-2-fluoro-5-iodo-3-methylbenzene

Precaución: for Research Use Only

Formula: C₇H₅ClFI
MW: 270.47 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28751663
CAS Number: 1807258-69-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR