tert-Butyl 5-bromo-3-chloro-2-fluorobenzoate ≥95%

Supplier: AOBChem USA

Warning

47258-500MG 47258-1G 47258-5G 47258-25G
77327-306EA 202.91 USD
77327-306 77350-890 77363-644 77386-164
tert-Butyl 5-bromo-3-chloro-2-fluorobenzoate ≥95%
tert-Butyl 5-bromo-3-chloro-2-fluorobenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₁BrClFO₂
Storage Temperature: Ambient
MDL Number: MFCD32707255
CAS Number: 2484888-82-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR