4-Chloro-1-(cyclohexylmethyl)-2-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

47247-500MG 47247-1G 47247-5G
77339-062EA 547.96 USD
77339-062 77350-888 77375-214
4-Chloro-1-(cyclohexylmethyl)-2-methylbenzene ≥97%
4-Chloro-1-(cyclohexylmethyl)-2-methylbenzene

Precaución: for Research Use Only

Formula: C₁₄H₁₉Cl
MW: 222.754 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29093149
CAS Number: 1369910-77-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR