1-Bromo-2-chloro-4-(cyclohexylmethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

47244-500MG 47244-1G 47244-5G
77339-058EA 547.96 USD
77339-058 77350-884 77375-210
1-Bromo-2-chloro-4-(cyclohexylmethyl)benzene ≥97%
1-Bromo-2-chloro-4-(cyclohexylmethyl)benzene

Precaución: for Research Use Only

Formula: C₁₃H₁₆BrCl
MW: 287.623 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29092501
CAS Number: 1369869-73-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR