3-Bromo-5-(cyclohexylmethyl)pyridine ≥95%

Supplier: AOBChem USA

Warning

47240-500MG 47240-1G 47240-5G
77339-056EA 547.96 USD
77339-056 77350-878 77375-204
3-Bromo-5-(cyclohexylmethyl)pyridine ≥95%
3-Bromo-5-(cyclohexylmethyl)pyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₆BrN
MW: 254.166 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707253
CAS Number: 2432848-74-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR