1-Bromo-4-(cyclohexylmethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

47238-500MG 47238-1G 47238-5G
77336-382EA 387.28 USD
77336-382 77350-876 77375-202
1-Bromo-4-(cyclohexylmethyl)benzene ≥97%
1-Bromo-4-(cyclohexylmethyl)benzene

Precaución: for Research Use Only

Formula: C₁₃H₁₇Br
MW: 253.178 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707251
CAS Number: 1785619-14-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR