1-Bromo-3-(cyclohexylmethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

47237-500MG 47237-1G 47237-5G
77336-380EA 387.28 USD
77336-380 77350-874 77375-200
1-Bromo-3-(cyclohexylmethyl)benzene ≥95%
1-Bromo-3-(cyclohexylmethyl)benzene

Precaución: for Research Use Only

Formula: C₁₃H₁₇Br
MW: 253.178 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707250
CAS Number: 98446-79-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR