3-Bromo-5-(1-methyl-3-(trifluoromethyl)-1H-pyrazol-5-yl)pyridine ≥97%
Supplier: AOBChem USA
|
Warning
|
47230-500MG
47230-1G
47230-5G
47230-25G
77327-302EA
160.68
USD
77327-302
77345-788
77363-642
77386-162
3-Bromo-5-(1-methyl-3-(trifluoromethyl)-1H-pyrazol-5-yl)pyridine ≥97%
3-Bromo-5-(1-methyl-3-(trifluoromethyl)-1H-pyrazol-5-yl)pyridine
Precaución: for Research Use Only
Formula:
C₁₀H₇BrF₃N₃ Storage Temperature: Ambient |
MDL Number:
MFCD32707247 CAS Number: 2432849-17-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...