5-(furan-2-yl)-1-methyl-3-(trifluoromethyl)-1H-pyrazole ≥95%

Supplier: AOBChem USA

Warning

47228-500MG 47228-1G 47228-5G 47228-25G
77327-298EA 119.48 USD
77327-298 77343-608 77363-638 77375-192
5-(furan-2-yl)-1-methyl-3-(trifluoromethyl)-1H-pyrazole ≥95%
5-(furan-2-yl)-1-methyl-3-(trifluoromethyl)-1H-pyrazole

Precaución: for Research Use Only

Formula: C₉H₇F₃N₂O
MW: 216.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25968674
CAS Number: 905844-08-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR