5-(2-Bromo-4-fluorophenyl)-1-methyl-3-(trifluoromethyl)-1H-pyrazole ≥97%

Supplier: AOBChem USA

Warning

47226-500MG 47226-1G 47226-5G
77337-084EA 407.88 USD
77337-084 77350-868 77375-190
5-(2-Bromo-4-fluorophenyl)-1-methyl-3-(trifluoromethyl)-1H-pyrazole ≥97%
5-(2-Bromo-4-fluorophenyl)-1-methyl-3-(trifluoromethyl)-1H-pyrazole

Precaución: for Research Use Only

Formula: C₁₁H₇BrF₄N₂
MW: 323.084 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707244
CAS Number: 2432848-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR