N'-(3-Bromo-5-(tert-butyl)benzylidene)-4-methylbenzenesulfonohydrazide ≥97%

Supplier: AOBChem USA

Warning

47139-500MG 47139-1G 47139-5G 47139-25G
77327-294EA 139.05 USD
77327-294 77344-664 77363-624 77386-158
N'-(3-Bromo-5-(tert-butyl)benzylidene)-4-methylbenzenesulfonohydrazide ≥97%
N'-(3-Bromo-5-(tert-butyl)benzylidene)-4-methylbenzenesulfonohydrazide

Precaución: for Research Use Only

Formula: C₁₈H₂₁BrN₂O₂S
MW: 409.341 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756352
CAS Number: 2764734-12-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR