N'-(5-Bromo-2-methylbenzylidene)-4-methylbenzenesulfonohydrazide ≥97%

Supplier: AOBChem USA

Warning

47134-1G 47134-5G 47134-25G
77341-318EA 82.4 USD
77341-318 77363-622 77375-174
N'-(5-Bromo-2-methylbenzylidene)-4-methylbenzenesulfonohydrazide ≥97%
N'-(5-Bromo-2-methylbenzylidene)-4-methylbenzenesulfonohydrazide

Precaución: for Research Use Only

Formula: C₁₅H₁₅BrN₂O₂S
MW: 367.261 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707237
CAS Number: 2432855-13-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR