N'-(3-Bromo-2-fluorobenzylidene)-4-methylbenzenesulfonohydrazide ≥97%

Supplier: AOBChem USA

Warning

47132-1G 47132-5G 47132-25G
77341-006EA 66.95 USD
77341-006 77363-620 77375-172
N'-(3-Bromo-2-fluorobenzylidene)-4-methylbenzenesulfonohydrazide ≥97%
N'-(3-Bromo-2-fluorobenzylidene)-4-methylbenzenesulfonohydrazide

Precaución: for Research Use Only

Formula: C₁₄H₁₂BrFN₂O₂S
MW: 371.225 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707236
CAS Number: 2432855-06-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR