2-Bromo-1-(methoxymethyl)-4-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

47091-500MG 47091-1G 47091-5G 47091-25G
77327-292EA 234.84 USD
77327-292 77350-862 77363-610 77386-156
2-Bromo-1-(methoxymethyl)-4-methylbenzene ≥95%
2-Bromo-1-(methoxymethyl)-4-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₁BrO
MW: 215.087 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22491773
CAS Number: 1824417-66-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR