N'-(2-Bromo-4-methylbenzylidene)-4-methylbenzenesulfonohydrazide ≥97%

Supplier: AOBChem USA

Warning

47090-1G 47090-5G 47090-25G
77341-276EA 80.34 USD
77341-276 77363-608 77375-162
N'-(2-Bromo-4-methylbenzylidene)-4-methylbenzenesulfonohydrazide ≥97%
N'-(2-Bromo-4-methylbenzylidene)-4-methylbenzenesulfonohydrazide

Precaución: for Research Use Only

Formula: C₁₅H₁₅BrN₂O₂S
MW: 367.261 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707231
CAS Number: 2082699-21-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR