N'-(4-Chloro-2-methylbenzylidene)-4-methylbenzenesulfonohydrazide ≥97%

Supplier: AOBChem USA

Warning

47085-1G 47085-5G 47085-25G
77341-004EA 66.95 USD
77341-004 77363-606 77375-160
N'-(4-Chloro-2-methylbenzylidene)-4-methylbenzenesulfonohydrazide ≥97%
N'-(4-Chloro-2-methylbenzylidene)-4-methylbenzenesulfonohydrazide

Precaución: for Research Use Only

Formula: C₁₅H₁₅ClN₂O₂S
MW: 322.81 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707230
CAS Number: 2432855-04-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR