N'-(4-Bromo-2-methylbenzylidene)-4-methylbenzenesulfonohydrazide ≥97%

Supplier: AOBChem USA

Warning

47083-1G 47083-5G 47083-25G
77340-684EA 60.77 USD
77340-684 77363-604 77375-158
N'-(4-Bromo-2-methylbenzylidene)-4-methylbenzenesulfonohydrazide ≥97%
N'-(4-Bromo-2-methylbenzylidene)-4-methylbenzenesulfonohydrazide

Precaución: for Research Use Only

Formula: C₁₅H₁₅BrN₂O₂S
MW: 367.261 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707229
CAS Number: 2432855-15-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR