2-Chloro-1,3-difluoro-4-(methoxymethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

47079-500MG 47079-1G 47079-5G 47079-25G
77327-290EA 98.88 USD
77327-290 77342-706 77363-602 77375-156
2-Chloro-1,3-difluoro-4-(methoxymethyl)benzene ≥97%
2-Chloro-1,3-difluoro-4-(methoxymethyl)benzene

Precaución: for Research Use Only

Formula: C₈H₇ClF₂O
MW: 192.59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707228
CAS Number: 2484888-75-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR