1-Bromo-3-chloro-2-(methoxymethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

47068-500MG 47068-1G 47068-5G 47068-25G
77327-288EA 120.51 USD
77327-288 77343-716 77363-598 77375-152
1-Bromo-3-chloro-2-(methoxymethyl)benzene ≥95%
1-Bromo-3-chloro-2-(methoxymethyl)benzene

Precaución: for Research Use Only

Formula: C₈H₈BrClO
MW: 235.506 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403844
CAS Number: 2605894-55-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR