2-(Benzyloxy)-3-bromo-5-methylphenol ≥97%

Supplier: AOBChem USA

Warning

47061-500MG 47061-1G 47061-5G 47061-25G
77327-286EA 119.48 USD
77327-286 77343-606 77363-596 77375-150
2-(Benzyloxy)-3-bromo-5-methylphenol ≥97%
2-(Benzyloxy)-3-bromo-5-methylphenol

Precaución: for Research Use Only

Formula: C₁₄H₁₃BrO₂
MW: 293.156 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707224
CAS Number: 2484889-04-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR