N'-(2-Chloro-4-fluorobenzylidene)-4-methylbenzenesulfonohydrazide ≥97%

Supplier: AOBChem USA

Warning

46990-1G 46990-5G 46990-25G
77341-274EA 80.34 USD
77341-274 77363-592 77375-146
N'-(2-Chloro-4-fluorobenzylidene)-4-methylbenzenesulfonohydrazide ≥97%
N'-(2-Chloro-4-fluorobenzylidene)-4-methylbenzenesulfonohydrazide

Precaución: for Research Use Only

Formula: C₁₄H₁₂ClFN₂O₂S
MW: 326.774 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707222
CAS Number: 2432855-14-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR