N'-(2,4-Difluorobenzylidene)-4-methylbenzenesulfonohydrazide ≥97%

Supplier: AOBChem USA

Warning

46985-1G 46985-5G 46985-25G
77341-002EA 66.95 USD
77341-002 77363-590 77375-144
N'-(2,4-Difluorobenzylidene)-4-methylbenzenesulfonohydrazide ≥97%
N'-(2,4-Difluorobenzylidene)-4-methylbenzenesulfonohydrazide

Precaución: for Research Use Only

Formula: C₁₄H₁₂F₂N₂O₂S
MW: 310.319 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10182980
CAS Number: 454675-05-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR