2-Bromo-1,5-dichloro-3-fluorobenzene ≥95%

Supplier: AOBChem USA

Warning

46968-500MG 46968-1G 46968-5G 46968-25G
77327-282EA 198.79 USD
77327-282 77350-860 77363-586 77386-154
2-Bromo-1,5-dichloro-3-fluorobenzene ≥95%
2-Bromo-1,5-dichloro-3-fluorobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrCl₂F
Storage Temperature: Ambient
MDL Number: MFCD16817660
CAS Number: 1242339-89-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR