(3-Bromo-2-(trifluoromethoxy)phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

46967-500MG 46967-1G 46967-5G
77327-280EA 340.93 USD
77327-280 77350-858 77375-140
(3-Bromo-2-(trifluoromethoxy)phenyl)methanol ≥95%
(3-Bromo-2-(trifluoromethoxy)phenyl)methanol

Precaución: for Research Use Only

Formula: C₈H₆BrF₃O₂
MW: 271.031 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756335
CAS Number: 2167935-60-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR