1-Bromo-3-(4-methoxybenzyl)benzene ≥95%

Supplier: AOBChem USA

Warning

46963-500MG 46963-1G 46963-5G
77337-858EA 449.08 USD
77337-858 77350-856 77375-138
1-Bromo-3-(4-methoxybenzyl)benzene ≥95%
1-Bromo-3-(4-methoxybenzyl)benzene

Precaución: for Research Use Only

Formula: C₁₄H₁₃BrO
Storage Temperature: Ambient
MDL Number: MFCD23379019
CAS Number: 53039-49-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR