1-Bromo-2-(2-methylbenzyl)benzene ≥97%

Supplier: AOBChem USA

Warning

46962-500MG 46962-1G 46962-5G
77337-082EA 407.88 USD
77337-082 77350-854 77375-136
1-Bromo-2-(2-methylbenzyl)benzene ≥97%
1-Bromo-2-(2-methylbenzyl)benzene

Precaución: for Research Use Only

Formula: C₁₄H₁₃Br
Storage Temperature: Ambient
MDL Number: MFCD32707219
CAS Number: 884-27-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR