1-Bromo-2-(4-fluorobenzyl)benzene ≥97%

Supplier: AOBChem USA

Warning

46961-500MG 46961-1G 46961-5G
77337-080EA 407.88 USD
77337-080 77350-852 77375-134
1-Bromo-2-(4-fluorobenzyl)benzene ≥97%
1-Bromo-2-(4-fluorobenzyl)benzene

Precaución: for Research Use Only

Formula: C₁₃H₁₀BrF
MW: 265.121 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22053210
CAS Number: 68355-79-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR