1-Bromo-2-(4-methoxybenzyl)benzene ≥95%

Supplier: AOBChem USA

Warning

46960-500MG 46960-1G 46960-5G
77338-216EA 469.68 USD
77338-216 77350-850 77375-132
1-Bromo-2-(4-methoxybenzyl)benzene ≥95%
1-Bromo-2-(4-methoxybenzyl)benzene

Precaución: for Research Use Only

Formula: C₁₄H₁₃BrO
Storage Temperature: Ambient
MDL Number: MFCD08694255
CAS Number: 68355-78-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR