2-Bromo-4-(4-methoxybenzyl)-1-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

46958-500MG 46958-1G 46958-5G
77327-278EA 347.11 USD
77327-278 77350-846 77375-128
2-Bromo-4-(4-methoxybenzyl)-1-methylbenzene ≥97%
2-Bromo-4-(4-methoxybenzyl)-1-methylbenzene

Precaución: for Research Use Only

Formula: C₁₅H₁₅BrO
MW: 291.183 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707217
CAS Number: 2432848-89-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR