1,2-Difluoro-4-(4-methoxybenzyl)benzene ≥95%

Supplier: AOBChem USA

Warning

46957-500MG 46957-1G 46957-5G
77339-116EA 552.08 USD
77339-116 77350-844 77375-126
1,2-Difluoro-4-(4-methoxybenzyl)benzene ≥95%
1,2-Difluoro-4-(4-methoxybenzyl)benzene

Precaución: for Research Use Only

Formula: C₁₄H₁₂F₂O
Storage Temperature: Ambient
MDL Number: MFCD32707216
CAS Number: 2432848-50-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR