N'-((2,6-Dichloropyridin-3-yl)methylene)-4-methylbenzenesulfonohydrazide ≥97%

Supplier: AOBChem USA

Warning

46956-500MG 46956-1G 46956-5G 46956-25G
77327-276EA 85.49 USD
77327-276 77342-244 77363-584 77375-124
N'-((2,6-Dichloropyridin-3-yl)methylene)-4-methylbenzenesulfonohydrazide ≥97%
N'-((2,6-Dichloropyridin-3-yl)methylene)-4-methylbenzenesulfonohydrazide

Precaución: for Research Use Only

Formula: C₁₃H₁₁Cl₂N₃O₂S
Storage Temperature: Ambient
MDL Number: MFCD26940390
CAS Number: 1820748-57-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR