2-Chloro-4-(methoxymethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

46921-500MG 46921-1G 46921-5G
77338-696EA 510.88 USD
77338-696 77350-842 77375-122
2-Chloro-4-(methoxymethyl)benzaldehyde ≥97%
2-Chloro-4-(methoxymethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉ClO₂
MW: 184.62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707215
CAS Number: 1911646-24-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR